Anti-CCR6 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX109647
89355-970EA 623.96 CAD
89355-970
Anti-CCR6 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CCR6 (chemokine (C-C motif) receptor 6)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CCR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR