Anti-IgA Goat Polyclonal Antibody

Supplier: GeneTex
GTX77090
89353-482EA 332.63 CAD
89353-482
Anti-IgA Goat Polyclonal Antibody
Antibodies
Goat anti-Rat IgA Polyclonal antibody

Recommended Starting Dilutions: ELISA (coating): Use at a dilution of 1:200-1:500. WB: Use at a dilution of 1:2,000-1:15,000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR