Anti-IgG Chicken Polyclonal Antibody

Supplier: GeneTex
GTX77335
89353-644EA 371.38 CAD
89353-644
Anti-IgG Chicken Polyclonal Antibody
Antibodies
In a direct ELISA against purified rat IgG 10 ug/ml as sample, conjugate diluted 1/50,000 produced an O.D.=1.0 in a 15 minute reaction with TMB as substrate. Optimal conjugate working dilutions should be determined for your particular assay conditions. Chickens were immunized with purified rat IgG in Freund's adjuvant. Hyperimmune eggs were collected and an IgY fraction was prepared. The resulting anti-IgG was immunoaffinity purified and conjugated with horseradish peroxidase (HRP).

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Chicken
Isotype: IgY
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR