Anti-IgG Goat Polyclonal Antibody

Supplier: GeneTex

GTX77339
89353-648EA 291.32 CAD
89353-648
Anti-IgG Goat Polyclonal Antibody
Antibodies
In a direct ELISA against purified rat IgG 10 ug/ml as sample, conjugate diluted 1/125,000 produced an O.D.=1.0 in a 15 minute reaction with TMB as substrate. Optimal conjugate working dilutions should be determined for your particular assay conditions. Goats were immunized with purified rat IgG in Freund's adjuvant. Anti IgG was immunoaffinity purified and conjugated with horseradish peroxidase (HRP)

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR