Anti-RAX Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX77859
89366-354EA 732.3 CAD
89366-354
Anti-RAX Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Antibody to RAX

WB: Suggested dilution at 0.2-5.0 ug/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.

Type: Primary
Antigen: RAX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR