Anti-RAX Rabbit Polyclonal Antibody

Supplier: Genetex

GTX77868
89366-362EA 732.3 CAD
89366-362
Anti-RAX Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to RAX

For WB: Use at a concentration of 2-8 ug/ml in 5% skim milk/PBS buffer. Not tested in other applications. Optimal dilutions should be determined by researcher.

Type: Primary
Antigen: RAX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR