Anti-RPB2 Rabbit Polyclonal Antibody

GTX10332
89349-958EA 483.06 CAD
89349-958
Anti-RPB2 Rabbit Polyclonal Antibody
This 140 kDa protein is the second largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. This subunit, in combination with at least two other polymerase subunits, forms a structure within the polymerase that maintains contact in the active site of the enzyme between the DNA template and the newly synthesized RNA. It is suggested that Rpb2 plays a role in enhancing transcription from prostate specific antigen genes. These genes may be linked to prostate cancer progression.

Type: Primary
Antigen: RPB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR