Anti-SCD Rabbit Polyclonal Antibody

Supplier: Genetex

GTX59867
89363-748EA 711.73 CAD
89363-748
Anti-SCD Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to SCD

Western blotting (0.5-4 µg/ml). However, the optimal conditions should be determined individually. Other applications have not been determined. The antibody recognizes a ~37 kDa SCD. Rat kidney tissue lysate can be used as a positive control.

Type: Primary
Antigen: SCD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR