Anti-SCP1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX15090
89367-030EA 909.56 CAD
89367-030
Anti-SCP1 Rabbit Polyclonal Antibody
Antibodies
The synaptonemal complex is a proteinaceous complex that apparently mediates synapses during the zygotene stage and then disintegrates. It is a complex structure that unites homologous chromosomes during the prophase stage of meiosis. It is the result of chromosome pairing interactions and not the cause of them.

IF: Use at a dilution of 1/100 - 1/750. IHC-P: Use at a dilution of 1/750. IHC-Fr: Use at a dilution of 1/750. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SCP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR