Anti-TPX2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX30690
89367-692EA 756.09 CAD
89367-692
Anti-TPX2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to TPX2 (Targeting protein for Xklp2)

This antibody may be used for Western blotting, immunoprecipitation, and immunofluorescence. A band at ~100 kDa can be detected by Western blot. Other applications have not been tested. Suggested working dilutions: * WB : 1:1,000 IP : 1:10~1:100. ICC/IF : 1:1,000.*The investigator should determine the optimal working dilution for a specific application.

Type: Primary
Antigen: TPX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR