Anti-TUBG1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX11321
89357-378EA 932.04 CAD
89357-378
Anti-TUBG1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to gamma Tubulin - Centrosome Marker

IF: Use at a dilution of 1/1000. This concentration was determined for staining of methanolacetone-fixed rat fibroblasts (Rat-1 cell line). WB: Use at a dilution of 1/1000 for whole cell extract of the human cell line HeLa and at a dilution of 1/5000 for Xenopus whole egg extract. Detects a band of approximately 48 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TUBG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR