Anti-VEGF Chicken Polyclonal Antibody

GTX14078
89367-268EA 666.36 CAD
89367-268
Anti-VEGF Chicken Polyclonal Antibody
VEGF (Vascular Endothelial Growth Factor) is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability inducing properties specific for the vascular endothelium. Of the four isoforms of VEGF, the smaller two, VEGF 165 and VEGF 121, are secreted proteins and act as diffusible agents, whereas the larger two (VEGF 189 and VEGF 206) remain cell associated.

Type: Primary
Antigen: VEGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR