Anti-VEGF Goat Polyclonal Antibody

Supplier: GeneTex
GTX74566
89365-472EA 942.03 CAD
89365-472
Anti-VEGF Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to VEGF

Recommended Starting Dilutions: ELISA: 0.5 - 2.0 ug/ml Functional Assays: 0.05 - 0.10 ug/ml Western Blotting: 0.1 - 0.2 ug/ml Immunohistology - Paraffin: 5.0 - 15.0 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: VEGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR