Anti-AFP Goat Polyclonal Antibody

Supplier: GeneTex

GTX77527
89353-764EA 330.69 CAD
89353-764
Anti-AFP Goat Polyclonal Antibody
Antibodies
Goat anti - Human Alpha - Fetoprotein

Type: Primary
Antigen: AFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR