Anti-ATR Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX10327
89350-760EA 519.16 CAD
89350-760
Anti-ATR Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to ATR

For WB: Use at a dilution of 0.033 - 0.067 ?g/ml. Detects a band of approximately 300 kDa (predicted molecular weight: 307 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: ATR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR