Anti-ATR Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX74209
89367-760EA 720.33 CAD
89367-760
Anti-ATR Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ATR

Recommended Starting Dilutions: Western Blotting: Max Dilution 1/2000 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: ATR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR