Anti-Abeta Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX82974
89366-762EA 599.97 CAD
89366-762
Anti-Abeta Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to beta Amyloid

For ELISA: Use at a dilution of 1:10,000. For WB: Use at a concentration of 0.5-1.0?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Abeta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR