Anti-Abeta Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX74474
89367-792EA 758.75 CAD
89367-792
Anti-Abeta Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to beta Amyloid (1-42)

Recommended Starting Dilutions: Western Blotting: 1/500 - 1/1000 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: Abeta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR