Anti-BID Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX10640
89354-668EA 1279.06 CAD
89354-668
Anti-BID Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Bid Cleavage Site

WB: Use at a concentration of 0.5 - 1.0 ?g/ml. Predicted molecular weight: 15 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: BID
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR