Anti-BNIP3L Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX28399
89362-752EA 781.14 CAD
89362-752
Anti-BNIP3L Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to BNIP3L

Western blot at 0.5 to 1 ?g/ml.

Type: Primary
Antigen: BNIP3L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR