Anti-CALCA Rabbit Polyclonal Antibody

GTX28553
89352-492EA 565.53 CAD
89352-492
Anti-CALCA Rabbit Polyclonal Antibody
Rabbit polyclonal to Calcitonin

Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CALCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR