Anti-CALCR Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX74439
89365-456EA 389.3 CAD
89365-456
Anti-CALCR Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Calcitonin Receptor

Recommended Starting Dilutions: ELISA: Max Dilution 1/10,000 Immunohistology - Frozen: 1/10 - 1/50 Western Blotting: Max Dilution Reducing conditions Immunohistology - Paraffin: 1/10 - 1/50 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CALCR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR