Anti-CASP1 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX27978
89362-706EA 741.9 CAD
89362-706
Anti-CASP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Caspase 1

Type: Primary
Antigen: CASP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR