Anti-ITGAE Mouse Monoclonal Antibody [clone: LF61]

Supplier: GeneTex

GTX74984
89365-658EA 755.76 CAD
89365-658
Anti-ITGAE Mouse Monoclonal Antibody [clone: LF61]
Antibodies
Mouse monoclonal antibody [LF61] to Integrin alpha E

Type: Primary
Antigen: ITGAE
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR