Anti-CD11C Mouse Monoclonal Antibody [clone: BU15]

Supplier: GeneTex

GTX75795
89365-908EA 579.45 CAD
89365-908
Anti-CD11C Mouse Monoclonal Antibody [clone: BU15]
Antibodies
Mouse monoclonal antibody [BU15] to CD11c

Recommended Starting Dilutions: Flow Cytometry: 1/10 - 1/50 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD11C
Clonality: Monoclonal
Clone: BU15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR