Anti-CD1A Mouse Monoclonal Antibody [clone: NA1/34-HLK]

Supplier: GeneTex

GTX76536
89366-134EA 565.47 CAD
89366-134
Anti-CD1A Mouse Monoclonal Antibody [clone: NA1/34-HLK]
Antibodies
Mouse monoclonal antibody [NA1/34-HLK] to CD1a

Recommended Starting Dilutions: Flow Cytometry: 1/50 - 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: NA1/34-HLK
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR