Anti-CD33 Mouse Monoclonal Antibody [clone: WM53]

Supplier: GeneTex
GTX74840
89365-596EA 434.37 CAD
89365-596
Anti-CD33 Mouse Monoclonal Antibody [clone: WM53]
Antibodies
Mouse monoclonal antibody [WM53] to CD33

Recommended Starting Dilutions: Flow Cytometry: 1/10 - 1/50 Immunohistology - Frozen: 1/50 - 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR