Anti-CR1 Mouse Monoclonal Antibody [clone: E11]

Supplier: GeneTex

GTX76302
89366-048EA 504.11 CAD
89366-048
Anti-CR1 Mouse Monoclonal Antibody [clone: E11]
Antibodies
Mouse monoclonal antibody [E11] to CD35

Type: Primary
Antigen: CR1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR