Anti-CD36 Mouse Monoclonal Antibody [clone: SM0]

Supplier: GeneTex

GTX76433
89353-400EA 755.76 CAD
89353-400
Anti-CD36 Mouse Monoclonal Antibody [clone: SM0]
Antibodies
Mouse monoclonal antibody [SM0] to CD36

Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: SM0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR