Anti-CD36 Mouse Monoclonal Antibody [clone: SMØ]

Supplier: GeneTex

GTX76436
89366-098EA 916.37 CAD
89366-098
Anti-CD36 Mouse Monoclonal Antibody [clone: SMØ]
Antibodies
Mouse monoclonal antibody [SM0] to CD36

Recommended Starting Dilutions: Flow Cytometry: 1/25 - 1/50 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR