Anti-CD4 Mouse Monoclonal Antibody [clone: 1F6]

GTX75369
89365-774EA 306.01 CAD
89365-774
Anti-CD4 Mouse Monoclonal Antibody [clone: 1F6]
The CD4 cell surface antigen is expressed by the helper-inducer subset of T lymphocytes and also by monocytes.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 1F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR