Anti-CD4 Mouse Monoclonal Antibody

GTX75368
89367-862EA 559.16 CAD
89367-862
Anti-CD4 Mouse Monoclonal Antibody
Recognises the human CD4 cell surface antigen, expressed by the helper-inducer subset of T lymphocytes and also by monocytes.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR