Anti-CD5 Mouse Monoclonal Antibody [clone: 4C7]

Supplier: GeneTex
GTX75371
89365-776EA 314.12 CAD
89365-776
Anti-CD5 Mouse Monoclonal Antibody [clone: 4C7]
Antibodies
Mouse monoclonal antibody [4C7] to CD5

Recommended Starting Dilutions: Western Blotting: 1/25 - 1/50 Immunohistology - Paraffin: 1/25 - 1/50 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 4C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR