Anti-COX2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX73852
89367-132EA 1233.77 CAD
89367-132
Anti-COX2 Rabbit Polyclonal Antibody
Antibodies
Polyclonal Antibody to COX2

This antibody may be diluted to a titer of 1:50-1:100 in an ABC method. However, depending on the staining conditions employed, we suggest that final dilution should be determined by the user. We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation conditions should be determined by the user. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining.

Type: Primary
Antigen: COX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR