Anti-CSK Goat Polyclonal Antibody

Supplier: GeneTex

GTX74502
89367-796EA 758.75 CAD
89367-796
Anti-CSK Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to CSK

Recommended Starting Dilutions: ELISA: Max Dilution >1/32,000 Western Blotting: 0.5 ug/ml - 2.0 ug/ml Immunohistology - Paraffin: 0.1 ug/ml - 0.3 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CSK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR