Anti-CYP2C8 Rabbit Polyclonal Antibody

GTX74418
89367-780EA 792.45 CAD
89367-780
Anti-CYP2C8 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Cytochrome P450 2C8

Recommended Starting Dilutions: Functional Assays: 0.1 - 5ul Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CYP2C8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR