Anti-CYP2C8 Rabbit Polyclonal Antibody

GTX110244
89356-240EA 278.13 CAD
89356-240
Anti-CYP2C8 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to CYP2C8 (cytochrome P450, family 2, subfamily C, polypeptide 8)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: CYP2C8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR