Anti-CK4 Mouse Monoclonal Antibody [clone: 6B10]

Supplier: GeneTex

GTX11215
89350-810EA 639.69 CAD
89350-810
Anti-CK4 Mouse Monoclonal Antibody [clone: 6B10]
Antibodies
Mouse monoclonal [6B10] to Cytokeratin 4

IF: Use at a dilution of 1/300. IHC-P: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Detects a band of approximately 59 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CK4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR