Anti-CK4 Mouse Monoclonal Antibody [clone: 6B10]

GTX73797
89353-280EA 815.48 CAD
89353-280
Anti-CK4 Mouse Monoclonal Antibody [clone: 6B10]
This antibody is specific to 59 kD protein. Cytokeratin 4 belongs to the type B (basic) subfamily of high molecular weight cytokeratins and exists in combination with cytokeratin 13. Cytokeratin 4 is primarily found in the non-cornifying squamous epithelium, i.e. superficial and intermediate epithelial cells of esophagus, ectocervix, tongue, vagina, larynx, pharynx, epiglottis, anus and cornea. Cytokeratin 4 may be detected in squamous cell carcinomas derived from several non-cornified epithelia.

Type: Primary
Antigen: CK4
Clonality: Monoclonal
Clone: 6B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR