Anti-DNMT1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX30364
89352-656EA 862.14 CAD
89352-656
Anti-DNMT1 Rabbit Polyclonal Antibody
Antibodies
Anti - DNMT1 polyclonal

Type: Primary
Antigen: DNMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR