Anti-CCL11 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX27197
89362-478EA 565.64 CAD
89362-478
Anti-CCL11 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Eotaxin

WB: Use at a dilution of 1/5000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CCL11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR