Anti-CCL11 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX74044
89365-370EA 624.35 CAD
89365-370
Anti-CCL11 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Eotaxin

For WB: Use at a dilution of 1:1,700 - 1:2,300. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CCL11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR