Anti-MAPK Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX13909
89359-902EA 742.11 CAD
89359-902
Anti-MAPK Rabbit Polyclonal Antibody
Antibodies
ERK1 is ubiquitously expressed in various tissues and cells. It is activated by phosphorylation of neighbouring serine and threonine residues by MEK1 and MEK2 in response to diverse stimuli such as growth factors and mitogens.

For WB: Use at a concentration of 1-3 ?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: MAPK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR