Anti-MAPK Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX27947
89352-318EA 754.55 CAD
89352-318
Anti-MAPK Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to ERK1

For IHC: Use at an assay dependent dilution. For IP: Use at an assay dependent dilution. For WB: Use at a dilution of 1:200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: MAPK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR