Anti-GST Goat Polyclonal Antibody

Supplier: GeneTex
GTX30511
89352-678EA 748.7 CAD
89352-678
Anti-GST Goat Polyclonal Antibody
Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Other
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR