Anti-IgG Goat Polyclonal Antibody

Supplier: GeneTex
GTX77199
89353-540EA 228.71 CAD
89353-540
Anti-IgG Goat Polyclonal Antibody
Antibodies
The affinity purified anti IgG h+l reactes with all subclasses of IgG and with all classes of guinea pig immunoglobulins by immunodiffusion and IEP techniques due to the presence of anti light chains. In a direct ELISA against purified guinea pig immunoglobulins, conjugate diluted to 1/30,000 produced an O.D.=1.0 in a 15 minute reaction with TMB as substrate. Optimal conjugate working dilutions should be determined for your particular assay conditions.. Goats were immunized with purified guinea pig IgG isolated from pooled normal guinea pig serum. Serum was collected, anti-IgG antibodies were immunoaffinity purified off an antigen containing immunoabsorbant and conjugated with horseradish peroxidase (HRP).

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR