Anti-HP Goat Polyclonal Antibody

Supplier: Genetex
GTX77237
89353-566EA 381.05 CAD
89353-566
Anti-HP Goat Polyclonal Antibody
Goat anti - Bovine Haptoglobin

Type: Primary
Antigen: HP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR