Anti-HDAC1 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX59755
89363-738EA 767.76 CAD
89363-738
Anti-HDAC1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to HDAC1

Western blotting (0.5-4 µg/ml), Immunoprecipitation (10-20 µg/ml), and Immunohistochemistry (10-20 µg/ml). However, the optimal conditions should be determined individually. The antibody detects the 62 kDa histone deacetylase 1. It does not cross-react with other HDAC proteins including HDAC2, 3, 4, 5, 6, 7, and 8.

Type: Primary
Antigen: HDAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR