Anti-IGG Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX22410
89351-802EA 673.26 CAD
89351-802
Anti-IGG Rabbit Polyclonal Antibody
Antibodies
This antibody may be diluted to a titer of 1:200-1:500 in an ABC method. We suggest an incubation period of 30 minutes at room temperature. Proteolytic treatment is required prior to immunostaining of formalin fixed paraffin embedded tissue sections.

Type: Primary
Antigen: IGG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR