Anti-IL1B Chicken Polyclonal Antibody

GTX82043
89368-040EA 642.96 CAD
89368-040
Anti-IL1B Chicken Polyclonal Antibody
Chicken polyclonal to Interleukin 1 beta

ELISA (only tested on recombinant protein): Affinity-purified anti-IL-1B IgY as primary antibody (0.2 ug/ml). and Rabbit anti-IgY-HRP as 2nd antibody.

Type: Primary
Antigen: IL1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR